missing translation for 'onlineSavingsMsg'
Learn More

8-Methylquinoline, 97+%

Product Code. 11362086
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362086 5 g 5g
11372086 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362086 Supplier Thermo Scientific Alfa Aesar Supplier No. L03288.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 611-32-5 | C10H9N | 143.19 g/mol

8-Methylquinoline has been used in preparation of osmium chloridophosphine complexes, as quinoline carbene tautomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Methylquinoline has been used in preparation of osmium chloridophosphine complexes, as quinoline carbene tautomers.

Solubility
Soluble in water 0.1-0.5 g/100 mL at 17°C.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-32-5
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00006810
InChI Key JRLTTZUODKEYDH-UHFFFAOYSA-N
Synonym quinoline, 8-methyl, 8-methyl-quinoline, ccris 408, 8-methyl, o-toluquinoline, 8-metyl quinoline, 8-methyl quinoline, pubchem5806, acmc-209mpf, dsstox_cid_888
PubChem CID 11910
ChEBI CHEBI:48984
IUPAC Name 8-methylquinoline
SMILES CC1=C2N=CC=CC2=CC=C1

Specifications

Melting Point -80°C
Density 1.074
Boiling Point 134°C to 136°C (25 mmHg)
Flash Point 105°C (221°F)
Odor Unpleasant
Refractive Index 1.615
Quantity 5 g
Beilstein 111340
Solubility Information Soluble in water 0.1-0.5g/100 mL at 17°C.
Formula Weight 143.19
Percent Purity ≥97%
Chemical Name or Material 8-Methylquinoline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.