missing translation for 'onlineSavingsMsg'
Learn More

8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride, 97%, BLD Pharm™

Product Code. 31004623
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
100mg
10g
1g
250mg
25g
5g
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31004623 100 g 100g
30989156 1 g 1g
30989155 100 mg 100mg
30993528 250 mg 250mg
30995536 10 g 10g
30991517 25 g 25g
30992788 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 31004623 Supplier BLD Pharm Supplier No. BD111458100g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 54745-74-3
Molecular Formula C6H12ClNO
MDL Number MFCD09800611
Quantity 100 g
SMILES C1(O2)CNCC2CC1.[H]Cl
Molecular Weight (g/mol) 149.62
Percent Purity 97%
Chemical Name or Material 8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.