Learn More
Mitutoyo™ 811 Series 20 to 90 Shore D Analogue Display Durometers
Product Code. 11771584
Encompass
Change view
Click to view available options
Dimensions (L x W x H):
60 x 30 x 195 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
11771584 60 x 30 x 195 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11771584 Supplier Mitutoyo™ Supplier No. 811333
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Specifications
Display Analog
Depth (Metric) 60 mm
Dimensions (L x W x H) 60 x 30 x 195 mm
For Use With (Application) Hard elastomers, plastics, hard rubber, ebonite, etc.
Hardness 20 to 90 Shore D