missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 9,10-Bis(phenylethynyl)-2-ethylanthracene, 98%

Product Code. 15460318
Change view
Click to view available options
Quantity:
1g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15460318 1g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15460318 Supplier Alfa Aesar™ Supplier No. H53470.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 53158-83-1
Molecular Formula C32H22
MDL Number MFCD00190223
Quantity 1g
Synonym 2-ethyl-9,10-bis phenylethynyl anthracene, 9,10-bis phenylethynyl-2-ethylanthracene, 2-ethyl-9,10-bis 2-phenylethynyl anthracene
InChI Key KXTNQYWZDLPDLS-UHFFFAOYSA-N
SMILES CCC1=CC=C2C(=C1)C(C#CC1=CC=CC=C1)=C1C=CC=CC1=C2C#CC1=CC=CC=C1
IUPAC Name 2-ethyl-9,10-bis(2-phenylethynyl)anthracene
Molecular Weight (g/mol) 406.53
PubChem CID 323129
Formula Weight 406.53
Percent Purity 98%
Chemical Name or Material 9,10-Bis(phenylethynyl)-2-ethylanthracene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.