missing translation for 'onlineSavingsMsg'
Learn More

9,9-Bis(methoxymethyl)-9H-fluorene, TRC

Product Code. 30136146
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136146 1 g 1g
30133100 250 mg 250mg
17961145 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136146 Supplier Toronto Research Chemicals Supplier No. TRCB4860401G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 182121-12-6
Molecular Weight (g/mol) 254.32
IUPAC Name 9,9-bis(methoxymethyl)fluorene

Specifications

Molecular Formula C17 H18 O2
Quantity 1 g
Synonym 9,9-Bis(methoxymethyl)-9H-fluorene, 9,9-Bis(methoxylmethyl) fluorine, 9,9-Bis(methoxymethyl)fluorene, 9,9-Di(methoxymethyl)fluorene, FP 1
InChI Formula InChI=1S/C17H18O2/c1-18-11-17(12-19-2)15-9-5-3-7-13(15)14-8-4-6-10-16(14)17/h3-10H,11-12H2,1-2H3
SMILES COCC1(COC)c2ccccc2c3ccccc13
Formula Weight 254.1307
Chemical Name or Material 9,9-Bis(methoxymethyl)fluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.