missing translation for 'onlineSavingsMsg'
Learn More

9,9-Dimethyl-N-phenyl-9H-fluoren-2-amine, TRC

Product Code. 30385112
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385112 100 mg 100mg
30376114 10 mg -
30371652 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385112 Supplier Toronto Research Chemicals Supplier No. TRCD460488100MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 355832-04-1
Molecular Formula C21H19N
Molecular Weight (g/mol) 285.38
SMILES CC1(C)c2ccccc2c3ccc(Nc4ccccc4)cc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H19N/c1-21(2)19-11-7-6-10-17(19)18-13-12-16(14-20(18)21)22-15-8-4-3-5-9-15/h3-14,22H,1-2H3
Formula Weight 285.1517
Chemical Name or Material 9,9-Dimethyl-N-phenyl-9H-fluoren-2-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.