missing translation for 'onlineSavingsMsg'
Learn More

9-Aminoacridine, TRC

Product Code. 30012007
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012007 100 mg 100mg
30138432 1 g 1g
30137499 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012007 Supplier Toronto Research Chemicals Supplier No. TRCA190880100MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 90-45-9
Molecular Formula C13 H10 N2
Molecular Weight (g/mol) 194.23
Synonym 9-Acridinamine, Acridine, 9-amino- (8CI), 10-Amino-5-azaanthracene, 9-Aminoacridine, 9AA, Aminacrin, Aminacrine, Izoacridina, Monacrin, NSC 13000, NSC 28747
IUPAC Name acridin-9-amine
SMILES Nc1c2ccccc2nc3ccccc13

Specifica

Quantity 100 mg
InChI Formula InChI=1S/C13H10N2/c14-13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)13/h1-8H,(H2,14,15)
Formula Weight 194.0844
Chemical Name or Material 9-Aminoacridine
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.