missing translation for 'onlineSavingsMsg'
Learn More

9-Chloro-11beta,17,21-trihydroxy-16beta-methylpregna-1,4-diene-3,20-dione (Beclometasone), Mikromol™

Product Code. 18080460
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18080460 25 mg 25mg
18043770 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18080460 Supplier Mikromol Supplier No. MM0199.030025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

9-Chloro-11beta,17,21-trihydroxy-16beta-methylpregna-1,4-diene-3,20-dione (Beclometasone)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Beclometasone
CAS 4419-39-0
Analyte or Component Names Beclometasone
Shipping Condition Room Temperature
Molecular Formula C22 H29 Cl O5
Certifications/Compliance ISO 9001
Synonym 9-Chloro-11beta,17,21-trihydroxy-16beta-methylpregna-1,4-diene-3,20-dione, Beclometasone, CCI 2332, 9-Chloro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione, Pregna-1,4-diene-3,20-dione, 9-chloro-11β,17,21-trihydroxy-16β-methyl- (7CI,8CI), (11β,16β)-9-Chloro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9-Chloro-11β,17,21-trihydroxy-16β-methyl-pregna-1,4-diene-3,20-dione, 9-Chloro-16β-methylprednisolone, 9α-Chloro-16β-methylprednisolone, Beclometasone, Beclomethasone, Becolvent
InChI Formula InChI=1S/C22H29ClO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15-,16-,17-,19-,20-,21-,22-/m0/s1
SMILES C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO
IUPAC Name (8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 408.92
Quantity 25 mg
Formula Weight 408.1704 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.