missing translation for 'onlineSavingsMsg'
Learn More

9-Methylacridine, 96%

Product Code. 15456567
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15456567

Brand: Thermo Scientific Alfa Aesar H33718.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 611-64-3 | C14H11N | 193.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-64-3
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
MDL Number MFCD00143523
InChI Key FLDRLXJNISEWNZ-UHFFFAOYSA-N
Synonym acridine, 9-methyl, 5-methylacridine, 9-methylakridin, 9-methylakridin czech, unii-1f3q4u9uis, 9-methyl acridine, 5-methylacridine european, 1f3q4u9uis, 9-methylacridin, 9-methyl-acridine
PubChem CID 11913
IUPAC Name 9-methylacridine
SMILES CC1=C2C=CC=CC2=NC2=CC=CC=C12

Specifications

Melting Point 117°C to 119°C
Quantity 250 mg
Formula Weight 193.24
Percent Purity 96%
Chemical Name or Material 9-Methylacridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.