Description
- Extremely lightweight safety spectacles with a modern design
- Supravision sapphire coating is scratch-resistant and chemical-resistant
- Individual temple inclination, and length adjustment for a personalized fit
- Simple lens replacement
- Soft temples
Specifications
Specifications
| Lens Type | Supravision Sapphire |
| Certifications/Compliance (Europe) | EN 166:2001, EN 172:1994 + A1:2000 + A2:2001 |
| Lens Tint | Brown |
| Frame Color | Black |
| Model | 9195 Sykper |
| Lens Material | Polycarbonate |
| UV Protection | UV 400 |