Description
- Sporty spectacles ensure optimum coverage of the entire eye area
- Innovative hinged side arm for a personalized fit
- Lightweight design weighs only 23 grams
- Metal-free
Supravision Excellence Coating
- Interior: Anti-fog
- Exterior: Scratch-resistant and chemical-resistant
- Lenses are easy to clean and less susceptible to soiling
Supravision Sapphire Coating
- Inside: Scratch-resistant and chemical-resistant
- Outside: Scratch-resistant and chemical-resistant
- Non-stick properties allow for easy removal of greasy/watery dirt
Specifications
Specifications
| Certifications/Compliance | EN 166:2001, EN 172:1994 + A1:2000 + A2:2001 |
| Lens Type | Supravision Excellence |
| Lens Coating | Anti-fog, Scratch-resistant |
| Filter Type | Sunglare |
| Lens Tint | Gray |
| Product Type | Ergonomic Safety Glasses |
| Frame Color | Gray |
| Model | 9199 X-Fit |
| Transmittance Value | 0.23 |
| Weight (Metric) | 23 g |
| Show More |