missing translation for 'onlineSavingsMsg'
Learn More

9H-Fluorene-4-carboxylic acid, 96%, Thermo Scientific™

Product Code. 10163191 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10163191 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10163191 Supplier Thermo Scientific Acros Supplier No. 334020010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6954-55-8
CAS Min % 95.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C14H10O2
MDL Number MFCD00001112
Quantity 1g
Beilstein 09, 690
Synonym fluorene-4-carboxylic acid, 4-fluorenecarboxylic acid, maybridge1_002107, 9h-fluorene-4-carboxylicacid, fluorene-4-carboxylic acid, technical grade
InChI Key WJNBLUOXTBTGMB-UHFFFAOYSA-N
SMILES C1C2=CC=CC=C2C3=C(C=CC=C31)C(=O)O
IUPAC Name 9H-fluorene-4-carboxylic acid
Molecular Weight (g/mol) 210.232
PubChem CID 81402
Formula Weight 210.23
Percent Purity ≥95%
Chemical Name or Material 9H-Fluorene-4-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.