missing translation for 'onlineSavingsMsg'
Learn More

cis-Diamminedichloroplatinum(II), Pt 64.5% min, Thermo Scientific™

Potent Anticancer Agent That Blocks DNA Synthesis.

Brand:  Thermo Scientific Alfa Aesar 010471.01

Product Code. 11345978

  • POA / 0.25g

Please to purchase this item. Need a web account? Register with us today!

Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

15663-27-1
Cl2H6N2Pt
Cisplatin
300.06
Yellow micro crystals
0.25 g
MFCD00011623
Soluble in DMF. Insoluble in most common solvents
Odorless
cis-Diamminedichloroplatinum(II), Potent anticancer agent that blocks DNA synthesis
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.