missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl terephthalate, 99%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar A13220.0E

Quantity 2500g

Product Code. 11490733

Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Dimethyl terephthalate is used in the Polyester Film (Audio/Video Tape,X-ray Film,Photo Film), Polyester Fiber, Pet Bottle, Polyester Adhesive, Engineering Plastics. DMT is volatile, it is an intermediate in some schemes for the recycling of PET.

Solubility
It is slightly soluble in water but soluble in hot alcohol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under ordinary conditions. Keep away from Strong oxidizing agents, Strong acids, Strong bases, Nitrates
TRUSTED_SUSTAINABILITY
Specifications

Specifications

120-61-6
288°C
C10H10O4
2500g
14,9162
It is slightly soluble in water but soluble in hot alcohol and ether.
COC(=O)C1=CC=C(C=C1)C(=O)OC
194.19
194.19
Dimethyl terephthalate
1.29
146°C (294°F)
MFCD00008440
1107185
dimethyl terephthalate, dimethyl p-phthalate, 1,4-benzenedicarboxylic acid, dimethyl ester, dimethyl p-benzenedicarboxylate, di-me terephthalate, dimethyl 4-phthalate, terephthalic acid, dimethyl ester, dimethyl 1,4-benzenedicarboxylate, terephthalic acid dimethyl ester, methyl p-methoxycarbonyl benzoate
WOZVHXUHUFLZGK-UHFFFAOYSA-N
1,4-dimethyl benzene-1,4-dicarboxylate
8441
99%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.