accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOOCH3

CAS RN 120-61-6

IUPAC Name: 1,4-dimethyl benzene-1,4-dicarboxylate
Synonyms: dimethyl terephthalate dimethylterephthalate
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: WOZVHXUHUFLZGK-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)C(=O)OC
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (4)
  • (1)
  • (1)
  • (1)