missing translation for 'onlineSavingsMsg'
Learn More

Antibiotic A23187, 98%, Thermo Scientific™

An antibiotic and calcium ionophore

Brand:  Thermo Scientific Alfa Aesar J63020.MA

Quantity 10mg
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Product Code. 15465289

  • ONLINE EXCLUSIVE PRICE
    207.00€
    186.65€ / 10mg
    Save 20.35€
    9% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
A23187 (Mixed Calcium-Magnesium Salt) is used in cell activation experiments when calcium dose-response data are not required. Ca-Mg ratio ∼ 1:1. it is widely used as a compound in cell activation. It is used as a highly selective Ca2+ ionophore. it is used in studies of intracellular Ca2+ mediated events. A23187 has been observed to induce cell activation, differentiation or proliferation. In vitro experimentation has shown A23187 to boost ROS generation and platelet fragmentation. A23187 is a widely used ionophore. It has the ability to increase the intracellular concentrations of cations and is widely used to increase the intracellular levels of calcium. Visualization of Synaptic Ca2+ Calmodulin-Dependent Protein Kinase II Activity in Living Neurons.

Solubility
Soluble in DMSO with warming or sonication. Also soluble in ethanol or ethyl acetate

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

52665-69-7
10mg
MFCD00151202,MFCD00151202
14,1639
Soluble in DMSO with warming or sonication; Also soluble in ethanol or ethyl acetate
CNC1=CC=C2OC(C[C@@H]3OC4(CC[C@@H]3C)O[C@H](C(C)C(=O)C3=CC=CN3)[C@H](C)C[C@H]4C)=NC2=C1C(O)=O
523.63
523.62
Antibiotic A23187
White to Yellow
C29H37N3O6
1096801
Calcimycin; Calcium ionophore A23187
HIYAVKIYRIFSCZ-CVXKHCKVSA-N
5-(methylamino)-2-{[(2S,3S,8S,9R,11R)-3,9,11-trimethyl-8-[1-oxo-1-(1H-pyrrol-2-yl)propan-2-yl]-1,7-dioxaspiro[5.5]undecan-2-yl]methyl}-1,3-benzoxazole-4-carboxylic acid
97043288
98%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.