accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 52665-69-7

CH3HNO(S)O(S)CH3O(S)(R)H3C(R)CH3H3CONHNOHO

CAS RN 52665-69-7

IUPAC Name: 5-(methylamino)-2-{[(2S,3S,8S,9R,11R)-3,9,11-trimethyl-8-[1-oxo-1-(1H-pyrrol-2-yl)propan-2-yl]-1,7-dioxaspiro[5.5]undecan-2-yl]methyl}-1,3-benzoxazole-4-carboxylic acid
Synonyms: 5-(methylamino)-2-{[(2S,3S,8S,9R,11R)-3,9,11-trimethyl-8-[1-oxo-1-(1H-pyrrol-2-yl)propan-2-yl]-1,7-dioxaspiro[5.5]undecan-2-yl]methyl}-1,3-benzoxazole-4-carboxylic acid
Molecular Weight (g/mol): 523.63
Molecular Formula: C29H37N3O6
InChi Key: HIYAVKIYRIFSCZ-CVXKHCKVSA-N
SMILES: CNC1=CC=C2OC(C[C@@H]3OC4(CC[C@@H]3C)O[C@H](C(C)C(=O)C3=CC=CN3)[C@H](C)C[C@H]4C)=NC2=C1C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)
Color 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results