missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ A23187, free acid

Description
Calcium ionophore that induces Ca2+-dependent cell death by increasing intracellular calcium concentration. Promotes intracelllular ROS generation and platelet particle formation (fragmentation) in vitro and in vivo. Can be used to induce autophagy in mammalian cells.
Specifications
Specifications
| CAS | 52665-69-7 |
| Quantity | 10 mg |
| Molecular Formula | C29H37N3O6 |
| Formula Weight | 523.63 |
| Purity | >98% |
| Molecular Weight (g/mol) | 523.63 |
| Chemical Name or Material | A23187, free acid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?