missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CLEC14A Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB7436T-100UG
This item is not returnable.
View return policy
Description
CLEC14A Monoclonal specifically detects CLEC14A in Human samples. It is validated for Flow Cytometry.
Specifications
| CLEC14A | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q86T13 | |
| CLEC14A | |
| Mouse myeloma cell line NS0-derived recombinant human CLEC14A Glu22-Ala397 Accession # Q86T13 | |
| RUO | |
| 161198 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 743940 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| C14orf27, CEG1, chromosome 14 open reading frame 27, ClECT and EGF-like domain containing protein, C-type lectin domain family 14 member A, C-type lectin domain family 14, member A, EGF R5, EGFR5, EGFR-5, Epidermal growth factor receptor 5 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CLEC14A in direct ELISA. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction