missing translation for 'onlineSavingsMsg'
Learn More

A-7 hydrochloride

Product Code. 15424479
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424479 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15424479 Supplier Thermo Scientific Alfa Aesar Supplier No. J62011.MB
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 79127-24-5 | C20H30Cl2N2O2S | 433.432 g/mol

Calmodulin antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 79127-24-5
Color White
Molecular Formula C20H30Cl2N2O2S
MDL Number MFCD00209822
Quantity 25 mg
Synonym a-7 hydrochloride, n-10-aminodecyl-5-chloro-1-naphthalenesulfonamide hydrochloride, n-10-aminodecyl-5-chloronaphthalene-1-sulfonamide hydrochloride, 1-10-aminodec-1-yl sulphamoyl-5-chloronaphthalene, n-10-aminodecyl-5-chloro-1-naphthalenesulphonamide hydrochloride, n-10-aminodecyl-5-chloronaphthalene-1-sulfonamide-hydrogen chloride 1/1
Solubility Information Soluble to 50mM in DMSO
InChI Key XDJCAQBTSCRBHS-UHFFFAOYSA-N
SMILES C1=CC2=C(C=CC=C2Cl)C(=C1)S(=O)(=O)NCCCCCCCCCCN.Cl
IUPAC Name N-(10-aminodecyl)-5-chloronaphthalene-1-sulfonamide;hydrochloride
Molecular Weight (g/mol) 433.432
PubChem CID 44119113
Formula Weight 433.44
Physical Form Solid
Chemical Name or Material A-7 hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.