missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ A.R.I.A Medium with 5% Horse Blood

Product Code. 15880110
Change view
Click to view available options
Description:
A.R.I.A Medium with 5% Horse Blood
Quantity:
10/Pk.
Manufacturing Origin:
United Kingdom
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity Manufacturing Origin Unit Size
15880110 A.R.I.A Medium with 5% Horse Blood 10/Pk. United Kingdom Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15880110 Supplier Thermo Scientific™ Supplier No. PB1243A
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate fastidious and non‑fastidious anaerobic bacteria using a general purpose anaerobic growth medium.

Isolate fastidious and non‑fastidious anaerobic bacteria from clinical samples within 24 to 72 hours using Thermo Scientific™ Anaerobe Recovery and Isolation Agar (A.R.I.A™).

Made from Thermo Scientific™ Oxoid™ dehydrated culture media, A.R.I.A contains 5% defibrinated horse blood and peptones, carefully selected to support the growth of anaerobic bacteria.

The following constituents have been added to the formulation to enhance performance:

  • Yeast extract provides a vitamin source, and starch is present to absorb any toxic metabolites
  • L-cysteine is present as a reducing agent and has shown to stimulate the growth of some anaerobes1
  • Glucose is the main carbohydrate, and sodium pyruvate is also added as an energy source for asaccharolytic cocci, such as Veillonella
  • Haemin and Vitamin K are also included as growth factors required by many Bacteriodes species2
TRUSTED_SUSTAINABILITY

Specifications

Color Ruby red
Content And Storage 2° to 10°C, away from light
Description A.R.I.A Medium with 5% Horse Blood
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar Plate
Fill Weight 18.5-20.5g
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.