missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A2LD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | A2LD1 |
|---|---|
| Concentration | 0.39 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16822275
|
Proteintech
23280-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16812275
|
Proteintech
23280-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene aids in the proteolytic degradation of crosslinked fibrin by breaking down isodipeptide L-gamma-glutamyl-L-epsilon-lysine, a byproduct of fibrin degradation. The reaction catalyzed by the encoded gamma-glutamylaminecyclotransferase produces 5-oxo-L-proline and a free alkylamine. Two transcript variants encoding the same protein have been found for this gene.Specifications
| A2LD1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q4KM86, Q923B0, Q9BVM4 | |
| 223267, 290500, 87769 | |
| A2LD1 Fusion Protein Ag18549 | |
| Primary | |
| -20°C | |
| GGACT |
| 0.39 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A2LD1, AIG2 like domain 1, GGACT | |
| Ggact | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title