missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AAGAB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546781
This item is not returnable.
View return policy
Description
Peptide sequence: LLNSLTGTNH SIGSADPCHP EQPHLPAADS TESLSDHRGG ASNTTDAQVD.
The protein encoded by this gene interacts with the gamma-adaptin and alpha-adaptin subunits of complexes involved in clathrin-coated vesicle trafficking. Mutations in this gene are associated with type I punctate palmoplantar keratoderma. Alternatively spliced transcript variants have been found for this gene.
Specifications
| AAGAB | |
| Polyclonal | |
| Unconjugated | |
| AAGAB | |
| 2310007F21Rik; AAGAB; alpha- and gamma-adaptin binding protein; alpha- and gamma-adaptin-binding protein p34; KPPP1; p34; PPKP1; PPKP1A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 79719 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6PD74 | |
| AAGAB | |
| Synthetic peptide directed towards the C-terminal of human AAGAB (aa 186-235). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction