missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AANAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526631
This item is not returnable.
View return policy
Description
PA5-26631 has successfully been used to detect serotonin N-AT in both WB and IF applications.
Arylalkylamine N-Acetyltransferase (AANAT) is the penultimate enzyme is melatonin synthesis. Hormonal melatonin has been implicated in sleep and circadian clock function. Large changes in the abundance of AANAT are believed to be responsible for the large day/night rhythms in melatonin synthesis in the pineal gland and retina. Most research shows that AANAT activity is regulated by controlling the steady-state levels of the protein. Recent research, however, suggests that another mechanism may also play a role in controlling melatonin production without altering AANAT protein and activity. This mechanism may explain the sudden physiological increase in circulating melatonin observed at the onset of the dark period in humans.
Specifications
| AANAT | |
| Polyclonal | |
| Unconjugated | |
| Aanat | |
| Aanat; AA-NAT; Aralkylamine N-acetyltransferase; arylakylamine N-acetyltransferase; Arylalkylamine N - acetyltransferase (Serotonin N - acetyltransferase); arylalkylamine N-acetyltransferase; DSPS; Nat-2; Nat4; Seretonin N-acetyltransferase; serotonin acetylase; Serotonin N-acetyltransferase; SNAT | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 15 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q16613 | |
| Aanat | |
| KLH conjugated synthetic peptide between 27-54 amino acids from the N-terminal region of human AANAT. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction