Learn More
AARE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14758-1-AP-150UL
 View more versions of this product
Product Code. 16853564
  • 410.00€ / 150µL
Description
This antibody can recognize at least 2 isoforms of APH.
This gene encodes the enzyme acylpeptide hydrolase, which catalyzes the hydrolysis of the terminal acetylated amino acid preferentially from small acetylated peptides. The acetyl amino acid formed by this hydrolase is further processed to acetate and a free amino acid by an aminoacylase. This gene is located within the same region of chromosome 3 (3p21) as the aminoacylase gene, and deletions at this locus are also associated with a decrease in aminoacylase activity. The acylpeptide hydrolase is a homotetrameric protein of 300 kDa with each subunit consisting of 732 amino acid residues. It can play an important role in destroying oxidatively damaged proteins in living cells. Deletions of this gene locus are found in various types of carcinomas, including small cell lung carcinoma and renal cell carcinoma.
Specifications
Specifications
AARE
Polyclonal
Unconjugated
APEH
AARE, ACPH, Acyl peptide hydrolase, Acylaminoacyl peptidase, APEH, APH, D3F15S2, D3S48E, DNF15S2, OPH, Oxidized protein hydrolase
Rabbit
Antigen Affinity Chromatography
RUO
235606, 24206, 327
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P13676, P13798, Q8R146
APEH
AARE Fusion Protein Ag6511
150 μL
Primary
Human, Mouse, Rat