Learn More
ATAGO™ Abbe Refractometer NAR-2T
Product Code. 11486238
Change view
Click to view available options
Dimensions (L x W x H):
120 x 200 x 250 mm (Refractometer), 100 x 110 x 70 mm (Thermometer)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
11486238 120 x 200 x 250 mm (Refractometer), 100 x 110 x 70 mm (Thermometer) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11486238 Supplier ATAGO™ Supplier No. 1220
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
The Abbe Refractometer NAR-2T is designed for use with compounds which require measurement at a high temperature.
  • Abbe table refractometer for the measurement of refractive index (1.3000 to 1.7000 ND) and Brix (0.0% to 95.0% scale) of high temperature samples between 5°C to 120°C and measures with thermometer unit from 0.0°C to 120°C.
  • LED illumination digital display of temperature readings in Celcius, and immediate analog scale display measurement readings with a D (deuterium) line (589 Nm) fiber optic light source
  • Operates on AC 100V to 240V, 50/60 Hz power supply, has a 110V power connection cord with a 6 ft. power cord, and can be calibrated with distilled water for ease of operation
  • Vertical, angled body (12 x 20 x 25 cm) with monocular, focusable, padded eyepiece for viewing
  • Can be connected to a constant temperature circulating bath (sold separately) for sample temperature control
Specifications
Type Benchtop Refractometer
Scale Range 0.0% to 95.0% (Brix)
Refractive Index 1.3000 to 1.7000
Accuracy ±0.0002 (Refractive Index), ±0.1% (Brix)
For Use With (Application) Measuring Compounds at High Temperatures
Weight (Metric) 2.8 kg (Refractometer), 0.5 kg (Thermometer)
Dimensions (L x W x H) 120 x 200 x 250 mm (Refractometer), 100 x 110 x 70 mm (Thermometer)
Temperature Range (Metric) 5°C to 120°C
Voltage 240 V