missing translation for 'onlineSavingsMsg'
Learn More

Abcam Human Fibronectin Antibody Pair - BSA and Azide free, 10 x 96 Tests

Product Code. 32112882 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 x 96 Tests
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32112882 10 x 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32112882 Supplier Abcam Supplier No. ab24183710x96Tests
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Antibody, suitable for sELISA.

  • Applications: sELISA
  • Species Reactivity: Human

Especificaciones

Applications Sandwich ELISA
Conjugate Unconjugated
Gene Symbols FN1
Quantity 10 x 96 Tests
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Secondary
Test Specificity This product reacts with Human.
Target Species Human
Product Type Secondary Antibody
Shipping Condition Blue Ice
Reactivity abrv Hu
Validated Application abrv sELISA
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.