missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG2/CD338 Antibody (5D3) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB995C
This item is not returnable.
View return policy
Description
ABCG2/CD338 Monoclonal antibody specifically detects ABCG2/CD338 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| ABCG2/CD338 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 5D3 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| ABC transporter, ABC15, ABCPMGC102821, ATP-binding cassette sub-family G member 2, ATP-binding cassette transporter G2, ATP-binding cassette, sub-family G (WHITE), member 2, BCRP1, BCRPMRX, BMDP, Breast cancer resistance protein, CD338, CD338 antigen, CDw338, EST157481, mitoxantrone resistance protein, Mitoxantrone resistance-associated protein, MXRMXR1, placenta specific MDR protein, Placenta-specific ATP-binding cassette transporter | |
| 3T3 cells transduced with human ABCG2 | |
| 0.1 mL | |
| ABC Transporters, Cancer, Cellular Markers, Hematopoietic Stem Cell Markers, Neuronal Cell Markers, Stem Cell Markers, Stem Cells | |
| 9429 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction