missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abiraterone N-Oxide, Mikromol
Description
Abiraterone N-Oxide, Mikromol
Specifications
Specifications
| Chemical Name or Material | Abiraterone N-Oxide |
| CAS | 2378463-76-2 |
| Analyte or Component Names | Abiraterone N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H31 N O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Androsta-5,16-dien-3-ol, 17-(1-oxido-3-pyridinyl)-, (3β)- (ACI), (3β)-17-(1-Oxido-3-pyridinyl)androsta-5,16-dien-3-ol (ACI) |
| InChI Formula | InChI=1S/C24H31NO2/c1-23-11-9-18(26)14-17(23)5-6-19-21-8-7-20(16-4-3-13-25(27)15-16)24(21,2)12-10-22(19)23/h3-5,7,13,15,18-19,21-22,26H,6,8-12,14H2,1-2H3/t18-,19-,21-,22-,23-,24+/m0/s1 |
| Solution Type | Neat |
| SMILES | C[C@]12CC[C@H]3[C@@H](CC=C4C[C@@H](O)CC[C@]34C)[C@@H]1CC=C2c5ccc[n+]([O-])c5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?