missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABLIM3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27981-1-AP-20UL
This item is not returnable.
View return policy
Description
The LIM domain is a double zinc finger structure that promotes protein-protein interactions. LIM domain proteins, such as ABLIM3, play roles in embryonic development, cell lineage determination, and cancer (Krupp et al., 2006 ).Specifications
| ABLIM3 | |
| Polyclonal | |
| Unconjugated | |
| ABLIM3 | |
| abLIM 3, ABLIM3, Actin binding LIM protein 3, HMFN1661, KIAA0843 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22885, 319713 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O94929, Q69ZX8 | |
| Ablim3 | |
| ABLIM3 Fusion Protein Ag27578 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction