missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ AbSelect™ Generic Antibody Purification Kit

Product Code. 18048685 Shop All Bio Techne Products
Change view
Click to view available options
Sufficient For:
1 Purification
3 Purifications
Unit Size:
1 test
3 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For unitSize
18048685 1 Purification 1 test
18005765 3 Purifications 3 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18048685 Supplier Novus Biologicals™ Supplier No. 8600005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fast and simple method to purify antibodies from ascites fluid and serum

The AbSelect method involves capture of the antibody on protein A resin and the removal of unwanted substances by a simple wash procedure. The purified product is then eluted and neutralized. The antibody to be purified or cleaned up ideally is in a volume of 100μL to 0.5mL, up to 500μg of antibody can be purified in each run.

Compatible with lightning-link antibody labeling system, which allows purified antibody to be rapidly conjugated with wide variety of labels

Recommended Storage

Store at 4°C. Do not freeze.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Antibody Purification Kit
Sufficient For 1 Purification
Content And Storage 4°C. Do not freeze.
Quantity 1 Test
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.