missing translation for 'onlineSavingsMsg'
Learn More

ABT1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-22332APC

Product Code. 30032152

  • 615.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ABT1 Polyclonal antibody specifically detects ABT1 in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ABT1
Polyclonal
Western Blot, Immunoprecipitation
activator of basal transcription 1, Basal transcriptional activator, hABT1, TATA-binding protein-binding protein
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Activator of Basal Transcription 1 using the numbering given in entry NP_037507.1 (GeneID 29777).
0.1 mL
Primary
Human
Purified
Western Blot, Immunoprecipitation
APC
PBS
Rabbit
Immunogen affinity purified
RUO
29777
Store at 4C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.