missing translation for 'onlineSavingsMsg'
Learn More

ACAA2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

220.00€ - 326.00€

Specifications

Antigen ACAA2
Dilution Western Blot 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:250-1:1000
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18657616
View Documents
Novus Biologicals
NBP3-06401-100ul
100 μg
326.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18620597
View Documents
Novus Biologicals
NBP3-06401-50ul
50 μg
220.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ACAA2 Polyclonal antibody specifically detects ACAA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

ACAA2
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cardiovascular Biology, Endocrinology, Signal Transduction
PBS, pH7.0
10449
IgG
Antigen and protein A Affinity-purified
Western Blot 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:250-1:1000
Polyclonal
Purified
RUO
Human
3-ketoacyl-CoA thiolase, mitochondrial, Acetyl-CoA acyltransferase, acetyl-CoA acyltransferase 2, acetyl-Coenzyme A acyltransferase 2, beta ketothiolase, beta-ketothiolase, DSAEC, EC 2.3.1, EC 2.3.1.16, FLJ35992, FLJ95265, Mitochondrial 3-oxoacyl-CoA thiolase, mitochondrial 3-oxoacyl-Coenzyme A thiolase, T1
Produced in rabbits immunized with E. coli-derived Human ACAA2 fragment.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.