missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACADS Mouse anti-Human, Clone: 8B10, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59496-50UL
This item is not returnable.
View return policy
Description
ACADS Monoclonal specifically detects ACADS in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| ACADS | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| ACAD3, acyl-CoA dehydrogenase, C-2 to C-3 short chain, acyl-Coenzyme A dehydrogenase, C-2 to C-3 short chain, Butyryl-CoA dehydrogenase, EC 1.3.99, EC 1.3.99.2, mitochondrial short-chain specific acyl-CoA dehydrogenase, SCADshort-chain acyl-CoA dehydrogenase, short-chain specific acyl-CoA dehydrogenase, mitochondrial, unsaturated acyl-CoA reductase | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 35 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 8B10 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| ACADS | |
| Recombinant human ACADS (25-412aa) purified from E. coli | |
| 50 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction