missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACADS Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | ACADS |
|---|---|
| Concentration | 0.24 mg/mL |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16896294
|
Proteintech
16623-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16886294
|
Proteintech
16623-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Introduces a double bond at position 2 in saturated acyl-CoA′s of short chain length, i.e. less than 6 carbon atoms.Specifications
| ACADS | |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Non-human Primate, Human, Rat, Mouse | |
| P15651, P16219, Q07417 | |
| 11409, 35, 64304 | |
| ACADS Fusion Protein Ag9922 | |
| Primary | |
| -20°C | |
| ACADS |
| 0.24 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| ACAD3, ACADS, Butyryl CoA dehydrogenase, SCAD | |
| ACADS | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title