missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACADSB Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99387B
This item is not returnable.
View return policy
Description
ACADSB Polyclonal antibody specifically detects ACADSB in Human samples. It is validated for Western Blot
Specifications
| ACADSB | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Biotin | |
| 2-MEBCAD, 2-methylbutyryl-coenzyme A dehydrogenase, ACAD7,2-methylbutyryl-CoA dehydrogenase, acyl-CoA dehydrogenase, short/branched chain, acyl-Coenzyme A dehydrogenase, short/branched chain, EC 1.3.99,2-methyl branched chain acyl-CoA dehydrogenase, EC 1.3.99.-, SBCADshort/branched chain specific acyl-CoA dehydrogenase, mitochondrial | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ACADSB. | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 36 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction