Learn More
Thermo Scientific™ Access Port with Cover (152 mm), Factory Install
Product Code. 15523761
Change view
Click to view available options
Diameter (Metric) Inside:
152 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside unitSize
15523761 152 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15523761 Supplier Thermo Scientific™ Supplier No. 193006
  • ONLINE EXCLUSIVE PRICE
    605.00€
    584.00€ / Pack of 1
    Save 21.00€
    3% Off
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
For Use With: Environmental Chambers and Incubators
Provide a point of entry for probes and monitoring devices in environmental chambers and incubators with Access Ports.
TRUSTED_SUSTAINABILITY
Specifications
Description Port with cover
Lighting LED
Type Access Port
Installation Factory
Diameter (Metric) Inside 152 mm