Learn More
Thermo Scientific™ Accessories for RadEye™ Radiation Detectors
Product Code. 16695295 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Docking Station
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
16695295 Docking Station Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16695295 Supplier Thermo Scientific™ Supplier No. 425067065
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Accessory for use with RadEye pocket-sized radiation detectors
  • Docking Station/Car Adapter includes charging circuitry (8-30 V DC, cigarette lighter plug), alarm relay, RS 232 interface, AAA NiMH rechargeable batteries
  • Goose neck mounting kit has suction cup for instanteneous attachment to windshield; fits to docking station (19-172-509) or desktop holder
  • Headset for RadEye includes earphone for RadEye series
TRUSTED_SUSTAINABILITY
Specifications
Product Type Docking Station