Learn More
Sartorius Accessories for Vivacell Models
Product Code. 10663382 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
For Vivacell 20 and 70; For Pressure-fuge head
Vivacell 250
For Use With (Application):
Charge Valve
Maintenance Kit
Includes:
Not Applicable
Sample Reservoir, Filtrate Container, O-ring Seals
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) For Use With (Application) Includes Unit Size
10663382 For Vivacell 20 and 70; For Pressure-fuge head Charge Valve Not Applicable Each
10059730 Vivacell 250 Maintenance Kit Sample Reservoir, Filtrate Container, O-ring Seals Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10663382 Supplier Sartorius Supplier No. VCA005
  • ONLINE EXCLUSIVE PRICE
    109.00€
    101.52€ / Pack of 1
    Save 7.48€
    6% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Accessories for various Vivacell units
  • Use charge value with pressure-fuge head
  • Maintenance Kit includes sample reservoir, filtrate container and 'O' ring seals
TRUSTED_SUSTAINABILITY
Specifications
Disposable Reusable
Filter Type Ultrafiltration
Sterility Non-sterile
For Use With (Application) Charge Valve
Includes Not Applicable
Type Accessory for Vivacell Model
For Use With (Equipment) For Vivacell 20 and 70; For Pressure-fuge head