Learn More
Thermo Scientific™ Accucore™ Polar Selectivity Kits
Verify optimum selectivity of Thermo Scientific™ Accucore™ HPLC columns for polar analytes.
Brand:  Thermo Scientific™ 17X26-052130-3VC
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 11367551
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
Use the Thermo Scientific™ Accucore™ Polar Selectivity Kits to verify optimum selectivity of Accucore HPLC columns for polar analytes.
Based on Core Enhanced Technology, Accucore columns provide fast, high-resolution separations—without the elevated backpressures required by sub-2μm particles.
  • Fast separations with excellent resolution and sensitivity
  • Lower backpressures so that UHPLC is not required
  • Kit contains 1 Accucore aQ HPLC Column, 1 Accucore PFP Column, and 1 Accucore HILIC Column
Specifications
Specifications
HPLC Column Kit
HPLC
Spherical Solid Core
80 Å
aQ C18, HILIC, PFP
Yes
2.1 mm
50 mm
2.6 μm
130 m²/g
Silica