Learn More
Testo™ Accurate Immersion/ Penetration Temperature Probe
Description
- The temperature probe featuring a long cable and flat tip is suitable for measurements in liquids and pastes as well as for air temperature measurements
- Thanks to its NTC sensor element, the probe provides precise temperature measurements with an accuracy of up to ±0.36 °F (±0.2 °C)
- The temperature probe features a 6 m cable. This exceptionally long cable means that it can also be used at measuring points located a long distance away from the Wi-Fi data logger.
- For long-term measurements, you can mount the temperature probe permanently at the measuring location. To do this you only need a clamp or PG gland (not included in delivery).
- The temperature probe is waterproof in accordance with protection class IP 67. Consequently, measurements can be taken outdoors for example.
- This probe's response time t99 (time it takes for the probe to register 99% of the jump in temperature) of 5 seconds refers to measurements in moving water at 140 °F (+60 °C). This response time is extended when, for example, measurements are taken in still liquid, pastes or air.
Specifications
Specifications
| IP Rating | IP67 |
| Accuracy | ±0.2°C (-25 to +75 °C), ±0.4 °C (Remaining Range) |
| Response Time | 5 sec. |
| Temperature Range (Metric) | -35°C to +80°C |
| Length (Metric) | 6 m |
| Shipping Weight (Metric) | 105 g |