missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACE-2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
199.00€
Specifications
| Antigen | ACE-2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
ACE-2 Polyclonal specifically detects ACE-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ACE-2 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Immunology, Virology Bacteria and Parasites | |
| PBS with 0.02% Sodium Azide | |
| ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15 | |
| ACE2 | |
| IgG | |
| Affinity Purified | |
| Anti-ACE2 has no cross response to ACE1. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| NP_068576 | |
| 59272 | |
| Antibody was raised against a peptide corresponding to 17 amino acids near the center of human ACE-2. The immunogen is located within amino acids 180 - 230 of ACE-2. Amino Acid Squence: RANHYEDYGDYWRGDYE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 90 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title