missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACE-2 Antibody (CL4035) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59037V
This item is not returnable.
View return policy
Description
ACE-2 Monoclonal antibody specifically detects ACE-2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| ACE-2 | |
| Monoclonal | |
| DyLight 405 | |
| ACEHangiotensin I converting enzyme 2, ACE-related carboxypeptidase, angiotensin I converting enzyme (peptidyl-dipeptidase A) 2, angiotensin-converting enzyme 2, Angiotensin-converting enzyme homolog, DKFZp434A014, EC 3.4.17, EC 3.4.17.23, Metalloprotease MPROT15 | |
| This antibody was developed using a recombinant protein derived from Q9BYF1, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Immunology, Virology Bacteria and Parasites | |
| 59272 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL4035 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction