missing translation for 'onlineSavingsMsg'
Learn More

Acebutolol EP Impurity C (Acebutolol USP Related Compound A), Mikromol™

Product Code. 30050709
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050709 100 mg 100mg
30050512 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050709 Supplier Mikromol Supplier No. MM0435.03
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Acebutolol EP Impurity C (Acebutolol USP Related Compound A)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(3-Acetyl-4-hydroxyphenyl)butanamide
CAS 40188-45-2
Analyte or Component Names N-(3-Acetyl-4-hydroxyphenyl)butanamide
Shipping Condition Room Temperature
Molecular Formula C12 H15 N O3
Certifications/Compliance ISO 17025
Synonym Acebutolol Hydrochloride Imp. C (EP), Acebutolol Imp. C (EP), Acebutolol USP Related Compound A, Acebutolol USP RC A, N-(3-Acetyl-4-hydroxyphenyl)butanamide, Acebutolol Related Compound A, Acebutolol Hydrochloride Impurity C, Acebutolol Impurity C
InChI Formula InChI=1S/C12H15NO3/c1-3-4-12(16)13-9-5-6-11(15)10(7-9)8(2)14/h5-7,15H,3-4H2,1-2H3,(H,13,16)
SMILES CCCC(=O)Nc1ccc(O)c(c1)C(=O)C
IUPAC Name N-(3-acetyl-4-hydroxyphenyl)butanamide
Molecular Weight (g/mol) 221.25
Quantity 100 mg
Formula Weight 221.1052 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.