missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACER2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575832
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The sphingolipid metabolite sphingosine-1-phosphate promotes cell proliferation and survival, whereas its precursor, sphingosine, has the opposite effect. The ceramidase ACER2 hydrolyzes very long chain ceramides to generate sphingosine.
Specifications
| ACER2 | |
| Polyclonal | |
| Unconjugated | |
| ACER2 | |
| 2410116I05Rik; ACER2; Acylsphingosine deacylase 3-like; alkaline CDase 2; alkaline ceramidase 2; AlkCDase 2; ALKCDase2; Asah3l; cancer related gene-liver 1 (CRG-L1); cancer-related gene liver 1 protein; ceramide hydrolase; CRG-L1; haCER2; maCER2; N-acylsphingosine amidohydrolase 3-like; PP11646 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 313339, 340485 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5QJU3 | |
| ACER2 | |
| Synthetic peptide corresponding to residues in Human ACER2. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction