missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial, ACS Reagent Grade, Ricca Chemical

Product Code. 31750429
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Amber Glass Bottle
Glass Bottle
Hard Pack
Natural Poly Bottle
Unit Size:
1 L
2.50 L
20 L
250mL
4 L
500mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31750429 500 mL Glass Bottle 500mL
31750107 250 mL Natural Poly Bottle 250mL
31750995 1 L Amber Glass Bottle 1 L
31749926 2.5 L Glass Bottle 2.50 L
31750006 4 L Amber Glass Bottle 4 L
31750224 20 L Hard Pack 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31750429 Supplier Ricca Chemical Company Supplier No. RABA0010500D1
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
Percent Purity 99.7% (min.)
Grade ACS
Quantity 500 mL
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation max. 0.001%
Chemical Name or Material Acetic Acid, Glacial
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetic acid, glacial, ACS Reagent Grade
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338+P310-IF IN EYES: Rinse gently with water for several minutes. Remove any contact lenses, if this can be done easily. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.