missing translation for 'onlineSavingsMsg'
Learn More

Acetone, ACS Reagent Grade, Ricca Chemical

Product Code. 31750794
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Metal Can
Natural Poly Bottle
Unit Size:
1 L
20 L
4 L
500mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31750794 4 L Natural Poly Bottle 4 L
31749606 500 mL Amber Glass Bottle 500mL
31750284 1 L Amber Glass Bottle 1 L
31751061 4 L Amber Glass Bottle 4 L
31750426 20 L Cubitainer 20 L
31750157 20 L Metal Can 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31750794 Supplier Ricca Chemical Company Supplier No. RSOA00104A
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Quantity 4 L
Packaging Natural Poly Bottle
Residue after Evaporation max. 0.001%
Titratable Acid max. 0.0003 meq/g
Titratable Base max. 0.0006 meq/g
Water max. 0.5%
Synonym acetone
Boiling Point 56°C
Percent Purity min. 99.5%
Grade ACS
Chemical Name or Material Acetone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.