missing translation for 'onlineSavingsMsg'
Learn More

Acetone E, Solstice

Product Code. 15985470
Change view
Click to view available options
Quantity:
7 L
Packaging:
Returnable Stainless Steel Can
Unit Size:
7 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15985470 7 L Returnable Stainless Steel Can 7 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15985470 Supplier Honeywell-Riedel-de Haen Supplier No. 344167L
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chromasolv™ for Pesticide Residue Analysis

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -95°C
CAS 67-64-1
Color Colorless
Quantity 7 L
Packaging Returnable Stainless Steel Can
Molecular Formula C3H6O
MDL Number MFCD00008765
Boiling Point 55°C to 57°C (1013 hPa)
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
Chemical Name or Material Acetone E
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.