missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Puriss. p.a., ACS Reagent, Reagent ISO, Reagent European Pharmacopoeia,Solstice

Product Code. 16615903
Change view
Click to view available options
Quantity:
195 L
Unit Size:
195 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16615903 195 L 195 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16615903 Supplier Fluka Supplier No. 65749195L
  • ONLINE EXCLUSIVE PRICE
    3005.00€
    2261.62€ / 195 L
    Save 743.38€
    24% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point -95°C
CAS 67-64-1
Color Colorless
Density 0.79 g/cm3 (20°C)
Quantity 195 L
Flash Point -17°C
Molecular Formula C3H6O
MDL Number MFCD00008765
Beilstein 635680
Synonym Acetonum
Boiling Point 55°C to 57°C (1013 hPa)
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
Grade ACS
Chemical Name or Material Acetone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.