missing translation for 'onlineSavingsMsg'
Learn More

Acetophenazine Maleate, Mikromol™

Product Code. 30049264
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049264 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049264 Supplier Mikromol Supplier No. MM1725.00
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Acetophenazine Maleate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetophenazine Maleate
CAS 5714-00-1
Analyte or Component Names Acetophenazine Maleate
Shipping Condition Room Temperature
Molecular Formula C23 H29 N3 O2 S . 2 C4 H4 O4
Certifications/Compliance ISO 9001
Synonym Ethanone, 1-[10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]-10H-phenothiazin-2-yl]-, (2Z)-2-butenedioate (1:2)Ethanone, 1-[10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]-10H-phenothiazin-2-yl]-, (2Z)-2-butenedioate (1:2) (salt) (9CI), Ethanone, 1-[10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]-10H-phenothiazin-2-yl]-, (Z)-2-butenedioate (1:2) (salt), Ketone, 10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]phenothiazin-2-yl methyl, dimaleate (6CI), Ketone, 10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]phenothiazin-2-yl methyl, maleate (1:2) (salt) (8CI), Ketone, 10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]phenothiazin-2-yl methyl, maleate (7CI), Acetophenazine dimaleate, Acetophenazine maleate, NSC 169180, NSC 70600, Phentoxate, Sch 6673, Tindal, Tindala
InChI Formula InChI=1S/C23H29N3O2S.2C4H4O4/c1-18(28)19-7-8-23-21(17-19)26(20-5-2-3-6-22(20)29-23)10-4-9-24-11-13-25(14-12-24)15-16-27;2*5-3(6)1-2-4(7)8/h2-3,5-8,17,27H,4,9-16H2,1H3;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1-
SMILES CC(=O)c1ccc2Sc3ccccc3N(CCCN4CCN(CCO)CC4)c2c1.OC(=O)\C=C/C(=O)O.OC(=O)\C=C/C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;1-[10-[3-[4-(2-hydroxyethyl)piperazin-1-yl]propyl]phenothiazin-2-yl]ethanone
Molecular Weight (g/mol) 643.7
Quantity 100 mg
Formula Weight 643.22 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.